Bismuth(III) nitrate basic, AnalaR NORMAPUR®
Plastic bottle for solids, 100 g
Supplier: VWR Chemicals
Bismuth(III) nitrate basic, AnalaR NORMAPUR®
Catalog # 84838.180
Supplier: VWR Chemicals
CAS Number: 1304-85-4
Jump to:
Product Details & Documents
Synonyms: Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
- CAS Number:1304-85-4
- Formula:Bi₅O(OH)₉(NO₃)₄
- MDL Number:MFCD00064844
- Molecular Weight:1461.99 g/mol
- Density:4.928 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-136-8
- UN:1477
- ADR:5.1,II
Documents
Certificates
Search by Lot Numberspec_VWRC84838.180 spec_VWRC84838.180 spec_VWRC84838.180
Product specification
Specifications
Specification Test Results
- Assay (calculated as Bi):71.0 - 74.0 %
- Identification:Passes test
- Loss on drying (105°C):Max. 3 %
- Substances not precipitated by ammonia:Max. 1 %
- Cl (Chloride):Max. 0.02 %
- Ag (Silver):Max. 25 ppm
- Cu (Copper):Max. 50 ppm
- Pb (Lead):Max. 20 ppm
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



