Order Entry
Belgium
ContactUsLinkComponent
2,3,5-Triphenyltetrazolium chloride (TTC) 5% in aqueous solution
2,3,5-Triphenyltetrazolium chloride (TTC) 5% in aqueous solution
Catalog # OXOISR0211C
CAS Number:  
2,3,5-Triphenyltetrazolium chloride (TTC) 5% in aqueous solution
Catalog # OXOISR0211C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :334.81 g/mol
EINECS:206-071-6
CAS Number:298-96-4
Formula:C₁₉H₁₅ClN₄
MDL Number:MFCD00011963