Order Entry
Belgium
ContactUsLinkComponent
Buffers, MES, 0,2 M solutions
Buffers, MES, 0,2 M solutions
Certificates
Buffers, MES, 0,2 M solutions
Certificates

Specifications

  • CAS No.:
    145224-94-8
  • MDL No.:
    MFCD00283848
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

Related Information

Frequently Bought Together