Order Entry
Belgium
ContactUsLinkComponent
 
Buffers, MES, 1,0 M solutions
 
Buffers, MES, 1,0 M solutions
 

 

  • CAS No.:
    145224-94-8
  • MDL No.:
    MFCD00283848
  • Physical form:
    Liquid
  • Appearance:
    Colourless

 

Frequently Bought Together