Order Entry
Belgium
ContactUsLinkComponent
TRItidy G™
TRItidy G™
Supplier:  PanReac AppliChem
Certificates
undefined
TRItidy G™
Supplier:  PanReac AppliChem

Specifications

  • Method / Format:
    Precipitation
    Organic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
    Total DNA
    Total protein
  • Sample type:
    Cultured cells
    Invertebrates
    Human/animal cells and tissues
  • Target molecule:
    DNA
    RNA
    Protein
    Co-purification of RNA, DNA and protein
  • Description:
    TRItidy G™

Specifications

Related Information

Frequently Bought Together