Order Entry
Belgium
ContactUsLinkComponent
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Inflammable
Santé
Irritant
Environnement
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Fournisseur:  Avantor
Certificats
undefined
Inflammable
Santé
Irritant
Environnement
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Fournisseur:  Avantor
Certificats

Résultats des tests de spécification

  • Assay (by GC):
    min. 99 %
  • Aromatic Compounds (as C6H6):
    max. 0.02 %
  • Boiling Range:
    max. 2 °C
  • Density (g/ml) at 25°C:
    max. 0.657
  • Recorded Boiling Point:
    68.7 °C
  • Residue after Evaporation:
    max. 5 ppm
  • Sulfur Compounds (as S):
    max. 0.005 %
  • Thiophene:
    passes test
  • Water (H2O):
    max. 0.02 %
  • Water Soluble Titrable Acid (meq/g):
    max. 0.0003
  • Aluminium (Al):
    max. 0.5
  • Boron (B):
    max. 0.02
  • Barium (Ba):
    max. 0.1
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.05
  • Cobalt (Co):
    max. 0.02
  • Chromium (Cr):
    max. 0.02
  • Copper (Cu):
    max. 0.02
  • Iron (Fe):
    max. 0.1
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.02
  • Nickel (Ni):
    max. 0.02
  • Lead (Pb):
    max. 0.1
  • Tin (Sn):
    max. 0.1
  • Zinc (Zn):
    max. 0.1

Résultats des tests de spécification

Informations associées

Frequently Bought Together