Order Entry
Belgium
ContactUsLinkComponent
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Assay:
    min. 99.9 % (v/v)
  • Acetone, isopropylalcohol:
    passes test
  • Color (APHA):
    max. 10
  • Methanol (CH3OH):
    max. 0.1 %
  • Residue after Evaporation:
    max. 0.001 %
  • Solubility in Water:
    passes test
  • Substances Darkened by H2SO4:
    passes test
  • Substances Reducing KMnO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0.0005
  • Titrable Base (meq/g):
    max. 0.0002
  • Water (H2O):
    max. 0.2 %
  • Aluminium (Al):
    max. 0.5
  • Boron (B):
    max. 0.02
  • Barium (Ba):
    max. 0.1
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.05
  • Cobalt (Co):
    max. 0.02
  • Chromium (Cr):
    max. 0.02
  • Copper (Cu):
    max. 0.2
  • Iron (Fe):
    max. 0.1
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.02
  • Nickel (Ni):
    max. 0.02
  • Lead (Pb):
    max. 0.1
  • Tin (Sn):
    max. 0.1
  • Zinc (Zn):
    max. 0.1

Specification Test Results

Related Information

Frequently Bought Together