Order Entry
Belgium
ContactUsLinkComponent
 
Flammable
Irritant
Ethyl acetate, BAKER ANALYZED® for LC-MS, J.T.Baker®
Catalog # 9828-03
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Ethyl acetate, BAKER ANALYZED® for LC-MS, J.T.Baker®
Catalog # 9828-03
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 - 400 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:
    ≤0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm:
    ≤0.050
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm:
    ≤1.00
  • Assay (CH₃COOC₂H₅):
    ≥99.6 %
  • Residue after Evaporation:
    ≤1.0 ppm
  • Water (H₂O):
    ≤0.05 %
  • LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):
    ≤50 ppb
  • Trace Impurities - Aluminum (Al), For Information Only:
    For Information Only
  • Trace Impurities - * Calcium (Ca), For Information Only:
    For Information Only
  • Trace Impurities - Iron (Fe), For Information Only:
    For Information Only
  • Trace Impurities - Magnesium (Mg), For Information Only:
    For Information Only
  • Trace Impurities - Potassium (K), For Information Only:
    For Information Only

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
 :GHS02
 :GHS07
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171