Microlab 600 Bubble Free Prime (BFP) syringes
The Bubble Free Prime (BFP) syringe is specifically designed for the Microlab® 600 series and the only syringe type compatible with the instrument. It has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
SaltLine syringes should be used when working with solutions that have a high salt concentration.
Frequently Bought Together