Order Entry
Belgium
Orders LinkContactUsLinkComponent
Diode lasers, class II

Diode lasers, class II

Supplier:   DIDALAB
Diode lasers, class II
Supplier:  DIDALAB
Certificates
Diode lasers, class II
Diode lasers, class II

2 Options Available Below

Product Details & Documents
  • Power slightly less than 1 mW
  • Homogenous and lightly divergent beam (3,2×7,1 mm at 10 metres)
  • Electronic device for constant power throughout its use
  • Polarized beam - ON / OFF switch

 : Laser provided on Ø 10 mm rod with mains power supply.

Product Family Options

Product Information

  • Wavelength
    Availability
    Price
  • DIDAPOD013133
    Diode laser, red, Class II
    Diode laser, red, Class II
    650 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,13 €
    125,13 €
    1 * 1 ST
       125,13 €

  • DIDAPOD013136
    Diode laser, blue, Class II
    Diode laser, blue, Class II
    405 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 341,25 €
    341,25 €
    1 * 1 ST
       341,25 €