ContactUsLinkComponent
Potassium hydroxide 0.5 M (0.5 N) in ethanol volumetric solution, Fluka™
Corrosive
Flammable
Potassium hydroxide 0.5 M (0.5 N) in ethanol volumetric solution, Fluka™
Certificates
Corrosive
Flammable
Potassium hydroxide 0.5 M (0.5 N) in ethanol volumetric solution, Fluka™
Certificates
Specification Test Results
  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective