Order Entry
Belgium
Orders LinkContactUsLinkComponent
Potassium nitrate 3 mol/l in aqueous solution

Potassium nitrate 3 mol/l in aqueous solution

Supplier:   Bernd Kraft
Potassium nitrate 3 mol/l in aqueous solution
Supplier:  Bernd Kraft
Certificates
Potassium nitrate 3 mol/l in aqueous solution

3 Options Available Below

Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-818-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    0,5 L
    Log in for availability
    Log in for availability
    1 * 0,5 L
    1 * 0,5 L

  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 76,70 €
    76,70 €
    1 * 1 L
       76,70 €

  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 194,00 €
    194,00 €
    1 * 5 L
       194,00 €

This product has been blocked by your organization. Please contact your purchasing department for more information
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.