Order Entry
Belgium
ContactUsLinkComponent
 
 
Sodium hydroxide 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
 :  Avantor
 
undefined
 
Sodium hydroxide 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
 :  Avantor
 

 

  • For Laboratory,Research,or Manufacturing Use:
    0.0995 - 0.1005
  • Protect from air to avoid absorption of carbon dioxide.:
    ≤5 ppm
  • Normality:
    0.0995 - 0.1005
  • Chloride (Cl):
    ≤5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm

 

Frequently Bought Together