Order Entry
Belgium
ContactUsLinkComponent
Dipotassium oxalate 300 g/l in aqueous solution
Irritant
Dipotassium oxalate 300 g/l in aqueous solution
Catalog # CMPLFC412791
CAS Number:  
Irritant
Dipotassium oxalate 300 g/l in aqueous solution
Catalog # CMPLFC412791
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :166.22 g/mol
Merck Index:12,07820
EINECS:209-506-8
 :GHS07
Storage Temperature:Ambient
CAS Number:583-52-8
Formula:C₂K₂O₄
MDL Number:MFCD00012464