Order Entry
Belgium
ContactUsLinkComponent
Perchloric acid 0.01 mol/l (0.01 N) in aqueous solution, Supelco®
Perchloric acid 0.01 mol/l (0.01 N) in aqueous solution, Supelco®
Catalog # 50439-1L
Supplier:  Merck
CAS Number:  
undefined
Perchloric acid 0.01 mol/l (0.01 N) in aqueous solution, Supelco®
Catalog # 50439-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :100.46 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-512-4
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325