Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.205 mol/l (0.205 N) in aqueous solution, AVS TITRINORM volumetric solution
Corrosive
Sodium hydroxide 0.205 mol/l (0.205 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30036.400
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide 0.205 mol/l (0.205 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30036.400
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.2046 - 0.2054 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:>105 °C (1013 hPa)
Density:1.008 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548