Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.3636 mol/l (0.3636 N) in aqueous solution
Irritant
Sodium hydroxide 0.3636 mol/l (0.3636 N) in aqueous solution
Catalog # 705-0063
Supplier:  Hach
CAS Number:  
Irritant
Sodium hydroxide 0.3636 mol/l (0.3636 N) in aqueous solution
Catalog # 705-0063
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.02 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
 :GHS07
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548