Order Entry
Belgium
ContactUsLinkComponent
 
 
Sodium hydroxide 0.25 mol/l (0.25 N), BAKER ANALYZED® volumetric solution, J.T.Baker®
  5638.9010
 :  Avantor
 :  
undefined
 
Sodium hydroxide 0.25 mol/l (0.25 N), BAKER ANALYZED® volumetric solution, J.T.Baker®
  5638.9010
 :  Avantor
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

 

 

  • Titer (mol/l):
    0.2485 - 0.2515
  • Chloride (Cl):
    max. 5
  • Iron (Fe):
    max. 0.5
  • Heavy Metals (as Pb):
    max. 1

 

 

 :
 :40 g/mol
 :∼105 °C (1013 hPa)
 :1.02 g/cm³ (25 °C)
 :Ambient
 :215-185-5
 :1824
 :8,III
 :GHS07
 :1310-73-2
 :NaOH
 :MFCD00003548