
Ethylene glycol, Technical Grade suitable as anti-freezing agent (with corrosion-inhibitor), blue
Catalog # 03770-5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Refractive Index:n20/D 1.431 (lit.)
- Appearance (color):complying
- Identity (IR):complying
- Refractive index (n 20/D):1.428 - 1.438
- Water (Karl Fischer):3 - 7 %
- Vapor Pressure:0.08 mmHg ( 20 °C)
- Vapor Density:2.1 (vs air)
- Explosion Limit:15.3 %
Specification Test Results
About this item
:
:62.07 g/mol
Merck Index:13,03832
Boiling Pt:196 - 198 °C (1013 hPa)
Melting Pt:-20 °C
Melting Pt:–20 °C
Density:1.115 g/cm³
Flash Pt:111 °C
Storage Temperature:Ambient
EINECS:203-473-3
REACH:01-2119456816-28
:GHS08
CAS Number:107-21-1
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885