Sodium thiosulphate 0.0109 mol/l (0.0109 N) in aqueous solution
Catalog # HACH2408949
Supplier: Hach
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
:
:158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499