Potassium chloride 3 mol/l (3 N) in aqueous solution
Catalog # WTWA50477006
Supplier: WTW
CAS Number:
Specifications
- Pk:6 Bottle
- Seq:0223
Specifications
About this item
:
:74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360