Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid ≥90.3% for testing of white vaseline acc. to DAB
Corrosive
Sulphuric acid ≥90.3% for testing of white vaseline acc. to DAB
Catalog # KRAF05702.3000
Supplier:  Bernd Kraft
CAS Number:  
Corrosive
Sulphuric acid ≥90.3% for testing of white vaseline acc. to DAB
Catalog # KRAF05702.3000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :98.08 g/mol
Density:1.82 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:1830
ADR:8,II
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589