Order Entry
Belgium
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
Catalog # AMBI9823
Supplier:  Ambion
CAS Number:  
undefined
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
Catalog # AMBI9823
Supplier:  Ambion
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :288.38 g/mol
Density:1…1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-788-1
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175