Order Entry
Belgium
ContactUsLinkComponent
Pig trypsin (from pancreas), MP Biomedicals
Pig trypsin (from pancreas), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Pig trypsin (from pancreas), MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    9002-07-7
  • Source:
    Pancreas
  • Species:
    Pig
  • Storage conditions:
    4 °C
  • Form:
    Off-white powder
  • MDL No.:
    MFCD00082094
  • Enzyme name:
    Trypsin
  • Enzyme activity:
    >250 U/mg
  • pH:
    ~8,0
  • Grade:
    Viirus-free, γ-irradiated

Specifications

Related Information

Frequently Bought Together