Order Entry
Belgium
ContactUsLinkComponent
Bromothymol blue 0.5 g/l in aqueous solution
Bromothymol blue 0.5 g/l in aqueous solution
Catalog # KRAF20682.0700
Supplier:  Bernd Kraft
CAS Number:  
Bromothymol blue 0.5 g/l in aqueous solution
Catalog # KRAF20682.0700
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :624.39 g/mol
Storage Temperature:Ambient
EINECS:200-971-2
CAS Number:76-59-5
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872