Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bromophenol blue in aqueous solution

Bromophenol blue in aqueous solution

Supplier:   Lovibond
Bromophenol blue in aqueous solution
Supplier:  Lovibond
Certificates
Bromophenol blue in aqueous solution

3 Options Available Below

Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 25,30 €
    25,30 €
    1 * 25 mL
       25,30 €

  • Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 36,50 €
    36,50 €
    1 * 100 mL
       36,50 €

  • Plastic bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 72,30 €
    72,30 €
    1 * 250 mL
       72,30 €

  • Plastic bottle
    500 mL
    Discontinued
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.