

Chloramine-T (sodium salt) 5% in aqueous solution analytical reagent
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:127-65-1
:
Formula:C₇H₇ClNNaO₂S
MDL Number:MFCD00000522
:227.65 g/mol
Storage Temperature:Ambient
EINECS:204-854-7
UN:3267
ADR:8,III
:GHS05
:GHS08
Frequently Bought Together