Order Entry
Belgium
ContactUsLinkComponent
Sodium hypochlorite 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Corrosive
Environment
Sodium hypochlorite 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Environment
Sodium hypochlorite 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance (Clear, light yellow liquid):
    Passes Test
  • Available Chlorine (when packaged):
    5.0 - 6.0 %
  • Phosphate (PO₄):
    ≤0.0005 %
  • Calcium (Ca):
    ≤0.001 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together