Order Entry
Belgium
ContactUsLinkComponent
Sodium hypochlorite (11 - 15% Cl₂)
Corrosive
Environment
Sodium hypochlorite (11 - 15% Cl₂)
Certificates
Corrosive
Environment
Sodium hypochlorite (11 - 15% Cl₂)
Certificates

Specification Test Results

  • Appearance (Colour) :
    Pale yellow to green
  • Assay (unspecified):
    10,5 - 15,5% available chlorine
  • Form:
    Liquid

Specification Test Results

Related Information

Frequently Bought Together