Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Corrosive
Potassium hydroxide concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4673.
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Potassium hydroxide concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4673.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • Normality (by titrimetry):
    Passes Test
  • (Makes 0.1N solution after dilution to 1000 mL):
  • Low in Carbonate:

Specification Test Results

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553