Order Entry
Belgium
ContactUsLinkComponent
 
EDTA disodium salt volumetric solution, for hardness of water determinations (1 ml = 1°dH/100 ml H₂O), Fluka™
Catalog # 34543-1L
CAS Number:  
EDTA disodium salt volumetric solution, for hardness of water determinations (1 ml = 1°dH/100 ml H₂O), Fluka™
Catalog # 34543-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :336.21 g/mol
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672