Order Entry
Belgium
ContactUsLinkComponent
Methyl orange 0.5 g/L in aqueous solution
Methyl orange 0.5 g/L in aqueous solution
Catalog # HACH14849
Supplier:  Hach
CAS Number:  
Methyl orange 0.5 g/L in aqueous solution
Catalog # HACH14849
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :327.34 g/mol
Storage Temperature:Ambient
EINECS:208-925-3
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502