Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide 0.01 mol/l (0.01 N) in ethanol
Potassium hydroxide 0.01 mol/l (0.01 N) in ethanol
Catalog # PRLSMC305253.1000
Supplier:  VWR Collection
CAS Number:  
Potassium hydroxide 0.01 mol/l (0.01 N) in ethanol
Catalog # PRLSMC305253.1000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553