Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Barium chloride
Barium chloride 24% in aqueous solution
Barium chloride 24% in aqueous solution
Barium chloride 24% in aqueous solution
PRLS89296.320
:
PROLABO CMP
:
Barium chloride 24% in aqueous solution
PRLS89296.320
:
PROLABO CMP
:
Pack type:
Glass bottle
Pk:
2,5 L
Seq:
5096
:
:
208.24 g/mol
:
∼100 °C (1013 hPa)
:
Ambient
:
233-788-1
:
3287
:
6.1,III
:
GHS07
:
10361-37-2
:
BaCl₂
:
MFCD00003445