Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Supplier: Thermo Fisher Scientific
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical Form: Liquid
MDL No.: MFCD00236358
2 Options Available Below
Product Details & Documents
Physical Form: Liquid
MDL No.: MFCD00236358
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J63487.K3
- J63487.K7
- Physical form:
- Liquid
- Liquid
- MDL No.:
- MFCD00236358
- MFCD00236358
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- J63487.K3TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)Glass bottle2 LLog in for availability1 * 2 L65,00 €Specifications:
More
Specifications
Cat. No.J63487.K3Physical formLiquidMDL No.MFCD00236358 - J63487.K7TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)Glass bottle4 LLog in for availability1 * 4 L102,00 €Specifications:
More
Specifications
Cat. No.J63487.K7Physical formLiquidMDL No.MFCD00236358
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



