

Potassium hydroxide 0.1 mol/l (0.1 N) Reag. Ph. Eur.
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:2924
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together