Phenolphthalein 1% in methanol
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:77-09-8
:
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913
:318.33 g/mol
Storage Temperature:Ambient
EINECS:201-004-7
UN:1230
ADR:3,II
:GHS02
:GHS06
:GHS08
Frequently Bought Together