Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-[(2,6-Dichlorobenzyl)oxy]-5-nitrobenzaldehyde
2-[(2,6-Dichlorobenzyl)oxy]-5-nitrobenzaldehyde
2-[(2,6-Dichlorobenzyl)oxy]-5-nitrobenzaldehyde
2-[(2,6-Dichlorobenzyl)oxy]-5-nitrobenzaldehyde
Catalog # MAYBBTB05370.1
Supplier:
Thermo Scientific
CAS Number:
Expand image
2-[(2,6-Dichlorobenzyl)oxy]-5-nitrobenzaldehyde
Catalog # MAYBBTB05370.1
Supplier:
Thermo Scientific
CAS Number:
Specifications
Pk:
1 g
Seq:
3047
Specifications
About this item
Product Details
:
CAS Number:
219539-46-5
Formula:
C₁₄H₉Cl₂NO₄
Product Details