3,5-Dimethyl-4-hydroxybenzonitrile ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4198-90-7
:
Formula:C₉H₉NO
MDL Number:MFCD00221716
:147.18 g/mol
:GHS07
Specification Test Results
- Purity (GC):min. 95.0 %
- Melting point:122.0 to 126.0 deg-C
Specification Test Results
Frequently Bought Together