3,5-Dimethyl-4-hydroxybenzonitrile
Catalog # MAYBRDP00227DA
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:147.18 g/mol
CAS Number:4198-90-7
Formula:C₉H₉NO
MDL Number:MFCD00221716