Order Entry
Belgium
ContactUsLinkComponent
3,5-Dimethyl-4-hydroxybenzonitrile
3,5-Dimethyl-4-hydroxybenzonitrile
Catalog # MAYB32-227.0001
Supplier:  Thermo Scientific
CAS Number:  
3,5-Dimethyl-4-hydroxybenzonitrile
Catalog # MAYB32-227.0001
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :147.18 g/mol
CAS Number:4198-90-7
Formula:C₉H₉NO
MDL Number:MFCD00221716