Sulphaquinoxaline
250 mg
Catalog # MOLE63005018-250MG
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 4-Amino-N-(quinoxalin-2-yl)benzenesulphonamide
- CAS Number:59-40-5
- Formula:C₁₄H₁₂N₄O₂S
- MDL Number:MFCD00055406
- Molecular Weight:300.3357 g/mol
- EINECS:200-423-2
Specifications
Specifications
- Pk:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



