Buffer, potassium phosphate, 0,2 M solution
Supplier: Thermo Fisher Scientific
About this item
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
8 Options Available Below
Product Details & Documents
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- J61261.AP
- J61261.K2
- J62397.AP
- J62397.K2
- J62239.AP
- J62239.K2
- J60851.AP
- J60851.K2
- Physical form:
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- Liquid
- CAS No.:
- 7778-77-0
- 7778-77-0
- 7778-77-0
- 7778-77-0
- 7778-77-0
- 7778-77-0
- 7778-77-0
- 7778-77-0
- MDL No.:
- MFCD00011401
- MFCD00011401
- MFCD00011401
- MFCD00011401
- MFCD00011401
- MFCD00011401
- MFCD00011401
- MFCD00011401
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- J61261.APPotassium phosphate, 0,2M buffer solution, pH 7,0Potassium phosphate, 0,2M buffer solution, pH 7,0Glass bottle500 mLLog in for availability1 * 500 mL96,50 €Specifications:
More
Specifications
Cat. No.J61261.APPhysical formLiquidCAS No.7778-77-0MDL No.MFCD00011401 - J61261.K2Potassium phosphate, 0,2M buffer solution, pH 7,0Potassium phosphate, 0,2M buffer solution, pH 7,0Glass bottle1 LLog in for availability1 * 1 L151,00 €Specifications:
More
Specifications
Cat. No.J61261.K2Physical formLiquidCAS No.7778-77-0MDL No.MFCD00011401 - J62397.APPotassium phosphate, 0,2M buffer solution, pH 7,4Potassium phosphate, 0,2M buffer solution, pH 7,4Glass bottle500 mLLog in for availability1 * 500 mL94,70 €Specifications:
More
Specifications
Cat. No.J62397.APPhysical formLiquidCAS No.7778-77-0MDL No.MFCD00011401 - J62397.K2Potassium phosphate, 0,2M buffer solution, pH 7,4Potassium phosphate, 0,2M buffer solution, pH 7,4Glass bottle1 LLog in for availability1 * 1 L147,00 €Specifications:
More
Specifications
Cat. No.J62397.K2Physical formLiquidCAS No.7778-77-0MDL No.MFCD00011401 - J62239.APPotassium phosphate, 0,2M buffer solution, pH 7,5Potassium phosphate, 0,2M buffer solution, pH 7,5Glass bottle500 mLLog in for availability1 * 500 mL88,00 €Specifications:
More
Specifications
Cat. No.J62239.APPhysical formLiquidCAS No.7778-77-0MDL No.MFCD00011401 - J62239.K2Potassium phosphate, 0,2M buffer solution, pH 7,5Potassium phosphate, 0,2M buffer solution, pH 7,5Glass bottle1 LLog in for availability1 * 1 L137,00 €Specifications:
More
Specifications
Cat. No.J62239.K2Physical formLiquidCAS No.7778-77-0MDL No.MFCD00011401 - J60851.APPotassium phosphate, 0,2M buffer solution, pH 8,0Potassium phosphate, 0,2M buffer solution, pH 8,0Glass bottle500 mLLog in for availability1 * 500 mL90,30 €Specifications:
More
Specifications
Cat. No.J60851.APPhysical formLiquidCAS No.7778-77-0MDL No.MFCD00011401 - J60851.K2Potassium phosphate, 0,2M buffer solution, pH 8,0Potassium phosphate, 0,2M buffer solution, pH 8,0Glass bottle1 LLog in for availability1 * 1 L140,00 €Specifications:
More
Specifications
Cat. No.J60851.K2Physical formLiquidCAS No.7778-77-0MDL No.MFCD00011401



