Order Entry
Belgium
ContactUsLinkComponent
Buffers, imidazole, 0,5 M solutions
Buffers, imidazole, 0,5 M solutions
Certificates
Buffers, imidazole, 0,5 M solutions
Certificates

Specifications

  • CAS No.:
    288-32-4
  • MDL No.:
    MFCD00005183
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together