Order Entry
Belgium
ContactUsLinkComponent
Buffer, EDTA buffer solution pH 8,5 (0,5 mol/L)
Buffer, EDTA buffer solution pH 8,5 (0,5 mol/L)
Catalog # R013
Supplier:  G-Biosciences
CAS Number:  
Buffer, EDTA buffer solution pH 8,5 (0,5 mol/L)
Catalog # R013
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    EDTA buffer solution pH 8,5 (0,5 mol/l)

Specifications

About this item

A DNase and RNase free solution of 0.5M EDTA.