Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, ACES, 0,5 M solution

Buffer, ACES, 0,5 M solution

Buffer, ACES, 0,5 M solution
Certificates
Buffer, ACES, 0,5 M solution

4 Options Available Below

Product Details & Documents

CAS No.: 7365-82-4
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00008030

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • J60366.AE
  • J60366.AK
  • J60755.AE
  • J60755.AK
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • CAS No.:
  • 7365-82-4
  • 7365-82-4
  • 7365-82-4
  • 7365-82-4
  • MDL No.:
  • MFCD00008030
  • MFCD00008030
  • MFCD00008030
  • MFCD00008030
  • Appearance:
  • Colourless
  • Colourless
  • Colourless
  • Colourless
  • Storage temperature:
  • 2°C to 8°C
  • 2°C to 8°C
  • 2°C to 8°C
  • 2°C to 8°C

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • J60366.AE
    ACES, 0,5M buffer solution, pH 6,5
    ACES, 0,5M buffer solution, pH 6,5
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 70,60 €
    70,60 €
    1 * 100 mL
       70,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60366.AE
    Physical form
    Liquid
    CAS No.
    7365-82-4
    MDL No.
    MFCD00008030
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • J60366.AK
    ACES, 0,5M buffer solution, pH 6,5
    ACES, 0,5M buffer solution, pH 6,5
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 105,00 €
    105,00 €
    1 * 250 mL
       105,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60366.AK
    Physical form
    Liquid
    CAS No.
    7365-82-4
    MDL No.
    MFCD00008030
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • J60755.AE
    ACES, 0,5M buffer solution, pH 7,0
    ACES, 0,5M buffer solution, pH 7,0
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 64,10 €
    64,10 €
    1 * 100 mL
       64,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60755.AE
    Physical form
    Liquid
    CAS No.
    7365-82-4
    MDL No.
    MFCD00008030
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C
  • J60755.AK
    ACES, 0,5M buffer solution, pH 7,0
    ACES, 0,5M buffer solution, pH 7,0
    Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 102,00 €
    102,00 €
    1 * 250 mL
       102,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    J60755.AK
    Physical form
    Liquid
    CAS No.
    7365-82-4
    MDL No.
    MFCD00008030
    Appearance
    Colourless
    Storage temperature
    2°C to 8°C