Order Entry
Belgium
ContactUsLinkComponent
 
Irritant
Flammable
Potassium hydroxide 0.1 M (0.1 N) in 2-propanol DIN 51558-1 volumetric solution, Fluka™
Catalog # 35008-1L
CAS Number:  
Irritant
Flammable
Potassium hydroxide 0.1 M (0.1 N) in 2-propanol DIN 51558-1 volumetric solution, Fluka™
Catalog # 35008-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.995 - 1.005
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :56.11 g/mol
Density:0.780 g/cm³
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:215-181-3
UN:1219
ADR:3,II
 :GHS07
 :GHS02
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553