Orders LinkContactUsLinkComponent
Cleanroom trousers, elasticated
Supplier:   FRISTADS KANSAS
Cleanroom trousers, elasticated
Supplier:  FRISTADS KANSAS
Certificates
Cleanroom trousers, elasticated
6 Options Available Below
Product Details & Documents
These trousers are made of XA32 fabric consisting of 99% polyester and 1% conductive fibres. The fibres are continuous filament fibres, which means that the garments do not emit particles or fibres. The carbon filaments produce good static discharge, thus preventing static charging and minimising triboelectric attraction of particles and fibres. This makes the garments very suitable for activities such as vehicle spraying.
  • Tracksuit-style
  • Elasticated waistband with drawstring
  • Open side legs
  • Inside key pocket
Weight: 125 g/m²
Specifications for Product Family
Specifications
  • Catalog No:
  • KWIN100631410XS
  • KWIN100631410S
  • KWIN100631410M
  • KWIN100631410L
  • KWIN100631410XL
  • KWIN1006314102XL
  • Model:
  • Elasticated waist
  • Elasticated waist
  • Elasticated waist
  • Elasticated waist
  • Elasticated waist
  • Elasticated waist
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Fabric:
  • 99% polyester and 1% conductive fibre
  • 99% polyester and 1% conductive fibre
  • 99% polyester and 1% conductive fibre
  • 99% polyester and 1% conductive fibre
  • 99% polyester and 1% conductive fibre
  • 99% polyester and 1% conductive fibre
  • Weight:
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
  • 125 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • Pale purple
    XS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,50 €
    65,50 €
    1 * 1 ST
       65,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    KWIN100631410XS
    Model
    Elasticated waist
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    99% polyester and 1% conductive fibre
    Weight
    125 g/m²
  • Pale purple
    S
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,50 €
    65,50 €
    1 * 1 ST
       65,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    KWIN100631410S
    Model
    Elasticated waist
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    99% polyester and 1% conductive fibre
    Weight
    125 g/m²
  • Pale purple
    M
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,50 €
    65,50 €
    1 * 1 ST
       65,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    KWIN100631410M
    Model
    Elasticated waist
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    99% polyester and 1% conductive fibre
    Weight
    125 g/m²
  • Pale purple
    L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,50 €
    65,50 €
    1 * 1 ST
       65,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    KWIN100631410L
    Model
    Elasticated waist
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    99% polyester and 1% conductive fibre
    Weight
    125 g/m²
  • Pale purple
    XL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,50 €
    65,50 €
    1 * 1 ST
       65,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    KWIN100631410XL
    Model
    Elasticated waist
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    99% polyester and 1% conductive fibre
    Weight
    125 g/m²
  • Pale purple
    XXL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,50 €
    65,50 €
    1 * 1 ST
       65,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    KWIN1006314102XL
    Model
    Elasticated waist
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Sleeve or leg length
    Long
    Fabric
    99% polyester and 1% conductive fibre
    Weight
    125 g/m²