Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Storage Bins

Storage Bins
 
Storage Bins
Storage Bins

 

 

These storage bins are particularly suitable for a wide range of applications in industry and craft.

  • Stable
  • Professional quality
  • Resistance to grease, resins, alkalis, oils and acids
Product Family Options

 

  • Colour
    W×D×H
     
     
  • Blue
    100×115×75 mm
       
       
    1 * 1 ST
    1 * 1 ST

  • Red
    100×115×75 mm
       
       
    1 * 24 ST
    1 * 24 ST - 48,80 €
    48,80 €
    1 * 24 ST
       48,80 €

  • Blue
    100×165×75 mm
       
       
    1 * 1 ST
    1 * 1 ST - 2,20 €
    2,20 €
    1 * 1 ST
       2,20 €

  • Red
    100×215×75 mm
       
       
    1 * 18 ST
    1 * 18 ST - 47,00 €
    47,00 €
    1 * 18 ST
       47,00 €

  • Orange
    100×215×75 mm
       
       
    1 * 18 ST
    1 * 18 ST

  • Blue
    145×240×125 mm
       
       
    1 * 1 ST
    1 * 1 ST - 4,70 €
    4,70 €
    1 * 1 ST
       4,70 €

  • Blue
    205×335×155 mm
       
       
    1 * 1 ST
    1 * 1 ST - 12,40 €
    12,40 €
    1 * 1 ST
       12,40 €

  • Blue
    315×495×185 mm
       
       
    1 * 1 ST
    1 * 1 ST - 28,90 €
    28,90 €
    1 * 1 ST
       28,90 €