Eurocontainers
Supplier: hünersdorff GmbH
Jump to:
About this item
These eurocontainers, made of polypropylene, without lids are ideal for storage and transport logistics. They are suitable for use in conveyor systems.
- High-quality plastic, resistant to customary alkalis and acids
- Closed, stable floor and side walls permit high stacking load capacity
- Impact-proof and fracture-proof and is 100% recyclable
- Food-safe and easy to clean
- Lightweight and high capacity
- Can be used in a temperature range of −40 to +90 °C
Ordering information: Individual lids for these containers can be ordered separately.
2 Options Available Below
Product Details & Documents
These eurocontainers, made of polypropylene, without lids are ideal for storage and transport logistics. They are suitable for use in conveyor systems.
- High-quality plastic, resistant to customary alkalis and acids
- Closed, stable floor and side walls permit high stacking load capacity
- Impact-proof and fracture-proof and is 100% recyclable
- Food-safe and easy to clean
- Lightweight and high capacity
- Can be used in a temperature range of −40 to +90 °C
Ordering information: Individual lids for these containers can be ordered separately.
Product Family Options
Product Information
- ColourTypeW×D×HMaterialAvailabilityPrice
- HUEN417400Eurocontainer, BasiclineEurocontainer, BasiclineLight greyWith 2 handles400×300×120 mmPPLog in for availability1 * 1 ST34,10 €
- HUEN417107Eurocontainer, SilverllineEurocontainer, SilverllineLight greyWith 2 handles600×400×420 mmPPLog in for availability1 * 1 ST120,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



